Assay Details
Gene Symbol: NBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111744.2 | XP_014967230.2 | 1375 | 92 | ||
XM_028831960.1 | XP_028687793.1 | 674 | 92 | |||
XM_028831961.1 | XP_028687794.1 | 1377 | 92 | |||
GenBank mRNA | JU334934.1 | - | 1347 | 92 | ||
JV063091.1 | - | 1042 | 92 | |||
JV716686.1 | - | 1365 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
