Assay Details
Gene Symbol: ELAPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194030.3 | NP_001180959.2 | 7 - 8 | 1185 | 88 | |
XM_028844953.1 | XP_028700786.1 | 1180 | 88 | |||
XM_028844954.1 | XP_028700787.1 | 1180 | 88 | |||
XM_028844955.1 | XP_028700788.1 | 1153 | 88 | |||
GenBank mRNA | JU321423.1 | - | 7 - 8 | 1123 | 88 | |
JV709601.1 | - | 7 - 8 | 943 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
