Assay Details
Gene Symbol: ELAPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194030.3 | NP_001180959.2 | 13 - 14 | 1915 | 101 | |
XM_028844953.1 | XP_028700786.1 | 1910 | 101 | |||
XM_028844954.1 | XP_028700787.1 | 1910 | 101 | |||
XM_028844955.1 | XP_028700788.1 | 1883 | 101 | |||
GenBank mRNA | JU321423.1 | - | 13 - 14 | 1853 | 101 | |
JV709601.1 | - | 13 - 14 | 1673 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
