Assay Details
Gene Symbol: TAF5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257498.1 | NP_001244427.1 | 3 - 4 | 308 | 61 | |
XM_015125718.2 | XP_014981204.1 | 359 | 61 | |||
XM_015125737.2 | XP_014981223.1 | 502 | 61 | |||
GenBank mRNA | JU333995.1 | - | 3 - 4 | 308 | 61 | |
JU475181.1 | - | 3 - 4 | 370 | 61 | ||
JV058003.1 | - | 3 - 4 | 391 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
