Assay Details
Gene Symbol: SAP30L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261790.1 | NP_001248719.1 | 419 | 115 | ||
GenBank mRNA | JU336406.1 | - | 670 | 115 | ||
JU477221.1 | - | 824 | 115 | |||
JV048261.1 | - | 419 | 115 | |||
JV636377.1 | - | 832 | 115 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
