Assay Details
Gene Symbol: ELAPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194030.3 | NP_001180959.2 | 21 - 22 | 3215 | 144 | |
GenBank mRNA | JU321423.1 | - | 21 - 22 | 3153 | 144 | |
JV709601.1 | - | 21 - 22 | 2973 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
