Assay Details
Gene Symbol: LRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260541.1 | NP_001247470.1 | 483 | 76 | ||
XM_028838404.1 | XP_028694237.1 | 667 | 76 | |||
XM_028838405.1 | XP_028694238.1 | 664 | 76 | |||
GenBank mRNA | JU326046.1 | - | 464 | 76 | ||
JV044861.1 | - | 483 | 76 | |||
JV616677.1 | - | 608 | 76 | |||
JV616678.1 | - | 608 | 76 | |||
JV667249.1 | - | 501 | 76 |
Gene Symbol: SLC7A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV714996.1 | - | 380 | 76 | ||
JV714997.1 | - | 380 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
