Assay Details
Gene Symbol: GABARAPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194264.2 | NP_001181193.1 | 147 | 61 | ||
GenBank mRNA | JU472337.1 | - | 94 | 61 | ||
JV045121.1 | - | 142 | 61 | |||
JV453052.1 | - | 155 | 61 | |||
JV635645.1 | - | 132 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194264.2 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
