Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002808223.3 | XP_002808269.2 | 4208 | 65 | ||
XM_015144632.2 | XP_015000118.1 | 4789 | 65 | |||
XM_015144633.2 | XP_015000119.1 | 4553 | 65 | |||
XM_015144635.2 | XP_015000121.1 | 4243 | 65 | |||
XM_015144637.2 | XP_015000123.1 | 4261 | 65 | |||
XM_015144640.2 | XP_015000126.1 | 5940 | 65 | |||
XM_015144644.2 | XP_015000130.1 | 4031 | 65 | |||
XM_015144646.2 | XP_015000132.1 | 538 | 65 | |||
XM_015144648.1 | XP_015000134.1 | 1022 | 65 | |||
XM_015144649.1 | XP_015000135.1 | 476 | 65 | |||
XM_015144652.2 | XP_015000138.1 | 483 | 65 | |||
XM_028831475.1 | XP_028687308.1 | 5986 | 65 | |||
XM_028831478.1 | XP_028687311.1 | 4263 | 65 | |||
XM_028831483.1 | XP_028687316.1 | 4034 | 65 | |||
XM_028831488.1 | XP_028687321.1 | 4666 | 65 | |||
GenBank mRNA | JU336667.1 | - | 5 - 6 | 3932 | 65 | |
JU471266.1 | - | 5 - 6 | 3932 | 65 | ||
JV048420.1 | - | 5 - 6 | 3903 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
