Assay Details
Gene Symbol: METTL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266641.1 | NP_001253570.1 | 926 | 70 | ||
GenBank mRNA | JU334314.1 | - | 924 | 70 | ||
JU334315.1 | - | 924 | 70 | |||
JU475419.1 | - | 926 | 70 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
