Assay Details
Gene Symbol: UBE2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194143.2 | NP_001181072.1 | 851 | 66 | ||
GenBank mRNA | JU472964.1 | - | 230 | 66 | ||
JV632821.1 | - | 233 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
