Assay Details
Gene Symbol: DAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194131.2 | NP_001181060.1 | 3 - 4 | 468 | 118 | |
XM_015143273.2 | XP_014998759.1 | 506 | 118 | |||
GenBank mRNA | JU320260.1 | - | 2 - 3 | 447 | 118 | |
JU473207.1 | - | 2 - 3 | 467 | 118 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
