Assay Details
Gene Symbol: C20H16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194229.2 | NP_001181158.1 | 640 | 97 | ||
GenBank mRNA | JU319631.1 | - | 593 | 97 | ||
JU475279.1 | - | 786 | 97 | |||
JV504087.1 | - | 594 | 97 | |||
JV579513.1 | - | 617 | 97 | |||
JV601007.1 | - | 896 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
