Assay Details
Gene Symbol: ATN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150940.2 | XP_015006426.1 | 3637 | 109 | ||
XM_015150942.2 | XP_015006428.1 | 3634 | 109 | |||
XM_028829221.1 | XP_028685054.1 | 3609 | 109 | |||
GenBank mRNA | JU474562.1 | - | 3571 | 109 | ||
JV046353.1 | - | 3571 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
