Assay Details
Gene Symbol: NUFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261811.1 | NP_001248740.1 | 2049 | 130 | ||
GenBank mRNA | JU335643.1 | - | 2106 | 130 | ||
JU476544.1 | - | 2049 | 130 | |||
JU476545.1 | - | 2049 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() |
