Assay Details
Gene Symbol: DHRS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194463.1 | NP_001181392.1 | 2 - 3 | 370 | 65 |
Gene Symbol: LOC114675548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842767.1 | XP_028698600.1 | 392 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
