Assay Details
Gene Symbol: ANKMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257747.1 | NP_001244676.1 | 1102 | 65 | ||
XM_015133908.2 | XP_014989394.1 | 963 | 65 | |||
XM_028844979.1 | XP_028700812.1 | 1124 | 65 | |||
GenBank mRNA | JU335559.1 | - | 1102 | 65 | ||
JU476482.1 | - | 1095 | 65 | |||
JV044439.1 | - | 1083 | 65 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
