Assay Details
Gene Symbol: DTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152877.2 | XP_015008363.1 | 1231 | 109 | ||
GenBank mRNA | JU324367.1 | - | 595 | 109 | ||
JV053793.1 | - | 601 | 109 | |||
JV644341.1 | - | 601 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
