Assay Details
Gene Symbol: RDH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257659.1 | NP_001244588.1 | 1 - 2 | 119 | 86 | |
GenBank mRNA | JU334786.1 | - | 1 - 2 | 119 | 86 | |
JU334787.1 | - | 1 - 2 | 119 | 86 | ||
JU334788.1 | - | 1 - 2 | 119 | 86 | ||
JU334789.1 | - | 1 - 2 | 119 | 86 | ||
JU334790.1 | - | 1 - 2 | 119 | 86 | ||
JU475824.1 | - | 1 - 2 | 119 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
