Assay Details
Gene Symbol: CDC42SE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257601.1 | NP_001244530.1 | 658 | 97 | ||
XM_015140702.2 | XP_014996188.1 | 685 | 97 | |||
XM_015140703.2 | XP_014996189.1 | 474 | 97 | |||
XM_015140705.2 | XP_014996191.1 | 495 | 97 | |||
GenBank mRNA | JU320135.1 | - | 487 | 97 | ||
JU320136.1 | - | 658 | 97 | |||
JU471686.1 | - | 642 | 97 | |||
JV043854.1 | - | 532 | 97 | |||
JV669230.1 | - | 670 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257601.1, XM_015140702.2, XM_015140703.2, XM_015140705.2 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
