Assay Details
Gene Symbol: CENPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091765.4 | XP_001091765.1 | 236 | 138 | ||
GenBank mRNA | JU491753.1 | - | 65 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() |
