Assay Details
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266922.1 | NP_001253851.1 | 4 - 5 | 587 | 123 | |
GenBank mRNA | JU336055.1 | - | 4 - 5 | 666 | 123 | |
JV048022.1 | - | 4 - 5 | 587 | 123 | ||
JV659564.1 | - | 4 - 5 | 671 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
