Assay Details
Gene Symbol: VRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112417.2 | XP_014967903.2 | 405 | 103 | ||
XM_028832465.1 | XP_028688298.1 | 405 | 103 | |||
XM_028832466.1 | XP_028688299.1 | 405 | 103 | |||
XM_028832467.1 | XP_028688300.1 | 405 | 103 | |||
GenBank mRNA | JU336249.1 | - | 325 | 103 | ||
JU336250.1 | - | 325 | 103 | |||
JU348056.1 | - | 325 | 103 |
Gene Symbol: FANCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU336251.1 | - | 325 | 103 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
