Assay Details
Gene Symbol: FCGRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839693.1 | XP_028695526.1 | 1063 | 107 | ||
XM_028839694.1 | XP_028695527.1 | 1036 | 107 | |||
XM_028839695.1 | XP_028695528.1 | 1031 | 107 | |||
XM_028839696.1 | XP_028695529.1 | 1007 | 107 | |||
XM_028839697.1 | XP_028695530.1 | 1037 | 107 | |||
XM_028839698.1 | XP_028695531.1 | 1083 | 107 |
Gene Symbol: LOC718859 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257520.1 | NP_001244449.1 | 743 | 107 | ||
GenBank mRNA | JU322969.1 | - | 743 | 107 | ||
JU474694.1 | - | 823 | 107 | |||
JV044676.1 | - | 929 | 107 | |||
JV044677.1 | - | 823 | 107 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
