Assay Details
Gene Symbol: MED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137413.2 | XP_014992899.1 | 937 | 72 | ||
XM_015137423.2 | XP_014992909.1 | 221 | 72 | |||
XM_028834896.1 | XP_028690729.1 | 937 | 72 | |||
XM_028834899.1 | XP_028690732.1 | 664 | 72 | |||
GenBank mRNA | JU472685.1 | - | 2 - 3 | 152 | 72 | |
JV641322.1 | - | 2 - 3 | 159 | 72 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_028834899.1 transcripts.
This assay is in the 5'UTR region of the XM_028834899.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
