Assay Details
Gene Symbol: SP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266828.1 | NP_001253757.1 | 2 - 3 | 143 | 111 | |
GenBank mRNA | JU336058.1 | - | 2 - 3 | 275 | 111 | |
JU476917.1 | - | 2 - 3 | 143 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
