Assay Details
Gene Symbol: OGG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260579.1 | NP_001247508.1 | 2651 | 69 |
Gene Symbol: CAMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257871.1 | NP_001244800.1 | 365 | 69 | ||
XM_015130262.2 | XP_014985748.2 | 430 | 69 | |||
GenBank mRNA | JU323226.1 | - | 2651 | 69 | ||
JU336824.1 | - | 365 | 69 | |||
JV048517.1 | - | 243 | 69 | |||
JV699453.1 | - | 399 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
