Assay Details
Gene Symbol: SPHK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120433.2 | XP_014975919.2 | 1196 | 66 | ||
XM_015120434.2 | XP_014975920.1 | 809 | 66 | |||
XM_015120435.2 | XP_014975921.1 | 1121 | 66 | |||
XM_015120436.2 | XP_014975922.1 | 1220 | 66 | |||
XM_015120437.2 | XP_014975923.1 | 1176 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
