Assay Details
Gene Symbol: FOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098399.1 | NP_001091869.1 | 3 - 4 | 504 | 67 | |
GenBank mRNA | AB219121.1 | - | 3 - 4 | 627 | 67 | |
EU073750.1 | - | 3 - 4 | 703 | 67 | ||
JU333721.1 | - | 3 - 4 | 648 | 67 | ||
JV601120.1 | - | 3 - 4 | 655 | 67 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
