Assay Details
Gene Symbol: TRIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111741.2 | XP_014967227.1 | 1995 | 65 | ||
XM_015111742.2 | XP_014967228.1 | 933 | 65 | |||
GenBank mRNA | JU320696.1 | - | 1068 | 65 | ||
JU475267.1 | - | 1963 | 65 | |||
JV044779.1 | - | 1982 | 65 | |||
JV636757.1 | - | 1983 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
