Assay Details
Gene Symbol: DDX39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047143.1 | NP_001040608.1 | 3 - 4 | 431 | 81 | |
XM_015135918.2 | XP_014991404.1 | 948 | 81 | |||
XM_015135919.2 | XP_014991405.1 | 924 | 81 | |||
GenBank mRNA | JU319866.1 | - | 4 - 5 | 463 | 81 | |
JU319867.1 | - | 4 - 5 | 669 | 81 | ||
JU472294.1 | - | 4 - 5 | 532 | 81 | ||
JU472295.1 | - | 4 - 5 | 662 | 81 | ||
JV048241.1 | - | 4 - 5 | 874 | 81 | ||
JV703364.1 | - | 4 - 5 | 668 | 81 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
