Assay Details
Gene Symbol: PIP4K2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015146607.2 | XP_015002093.1 | 497 | 103 | ||
XM_015146608.2 | XP_015002094.2 | 122 | 103 | |||
XM_015146609.2 | XP_015002095.1 | 492 | 103 | |||
GenBank mRNA | JU335005.1 | - | 2 - 3 | 346 | 103 | |
JU335006.1 | - | 2 - 3 | 346 | 103 | ||
JU335007.1 | - | 2 - 3 | 346 | 103 | ||
JU335008.1 | - | 2 - 3 | 346 | 103 | ||
JU335009.1 | - | 2 - 3 | 346 | 103 | ||
JU335010.1 | - | 2 - 3 | 346 | 103 | ||
JU476015.1 | - | 2 - 3 | 321 | 103 | ||
JV044228.1 | - | 2 - 3 | 294 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
