Assay Details
Gene Symbol: BCL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112435.2 | XP_014967921.1 | 426 | 72 | ||
XM_015112437.2 | XP_014967923.1 | 451 | 72 | |||
XM_015112438.2 | XP_014967924.1 | 461 | 72 | |||
XM_015112440.2 | XP_014967926.1 | 457 | 72 | |||
XM_015112443.2 | XP_014967929.1 | 446 | 72 | |||
XM_015112444.2 | XP_014967930.1 | 299 | 72 | |||
GenBank mRNA | JU335020.1 | - | 79 | 72 | ||
JU472470.1 | - | 236 | 72 | |||
JU476023.1 | - | 236 | 72 | |||
JV548031.1 | - | 73 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
