Assay Details
Gene Symbol: BLVRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258144.1 | NP_001245073.1 | 4 - 5 | 292 | 72 | |
XM_015133519.2 | XP_014989005.1 | 296 | 72 | |||
GenBank mRNA | JU322004.1 | - | 4 - 5 | 338 | 72 | |
JU472192.1 | - | 4 - 5 | 292 | 72 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
