Assay Details
Gene Symbol: PPP3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112591.2 | XP_014968077.1 | 915 | 100 | ||
XM_028832436.1 | XP_028688269.1 | 828 | 100 | |||
XM_028832437.1 | XP_028688270.1 | 656 | 100 | |||
GenBank mRNA | JU320071.1 | - | 5 - 6 | 843 | 100 | |
JU320072.1 | - | 5 - 6 | 851 | 100 | ||
JU320073.1 | - | 5 - 6 | 843 | 100 | ||
JU320074.1 | - | 5 - 6 | 843 | 100 | ||
JU473272.1 | - | 5 - 6 | 781 | 100 | ||
JV045353.1 | - | 5 - 6 | 852 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
