Assay Details
Gene Symbol: ECPAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015117108.2 | XP_014972594.2 | 3662 | 83 | ||
XM_015117110.2 | XP_014972596.2 | 3786 | 83 | |||
XM_028834988.1 | XP_028690821.1 | 3675 | 83 | |||
XM_028834989.1 | XP_028690822.1 | 3870 | 83 | |||
GenBank mRNA | JU341113.1 | - | 3509 | 83 | ||
JV710776.1 | - | 3510 | 83 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
