Assay Details
Gene Symbol: PFDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260521.1 | NP_001247450.1 | 2 - 3 | 177 | 72 | |
GenBank mRNA | JV043603.1 | - | 2 - 3 | 177 | 72 | |
JV601814.1 | - | 2 - 3 | 197 | 72 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
