Assay Details
Gene Symbol: KANSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151579.2 | XP_015007065.1 | 1332 | 83 | ||
XM_015151580.2 | XP_015007066.1 | 1464 | 83 | |||
XM_015151584.2 | XP_015007070.1 | 1528 | 83 | |||
XM_015151585.1 | XP_015007071.1 | 1506 | 83 | |||
GenBank mRNA | JU322110.1 | - | 1277 | 83 | ||
JU470535.1 | - | 1272 | 83 | |||
JV044367.1 | - | 1278 | 83 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
