Assay Details
Gene Symbol: SLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090273.4 | XP_001090273.2 | 2197 | 98 | ||
XM_028850070.1 | XP_028705903.1 | 2197 | 98 | |||
XM_028850071.1 | XP_028705904.1 | 2143 | 98 | |||
XM_028850072.1 | XP_028705905.1 | 2197 | 98 | |||
XM_028850073.1 | XP_028705906.1 | 2047 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
