Assay Details
Gene Symbol: EPM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087462.4 | XP_001087462.1 | 657 | 124 | ||
XM_028847191.1 | XP_028703024.1 | 384 | 124 | |||
GenBank mRNA | JU326005.1 | - | 1 - 2 | 233 | 124 | |
JU326006.1 | - | 1 - 2 | 233 | 124 | ||
JV557740.1 | - | 1 - 2 | 276 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
