Assay Details
Gene Symbol: PML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042434.1 | NP_001035899.1 | 1708 | 65 | ||
XM_015142614.2 | XP_014998100.1 | 1699 | 65 | |||
XM_015142617.2 | XP_014998103.1 | 1843 | 65 | |||
XM_015142619.2 | XP_014998105.1 | 1851 | 65 | |||
XM_015142622.2 | XP_014998108.1 | 1707 | 65 | |||
GenBank mRNA | DQ231471.1 | - | 1708 | 65 | ||
JU323536.1 | - | 1686 | 65 | |||
JU323852.1 | - | 1830 | 65 | |||
JU476034.1 | - | 1648 | 65 | |||
JU476035.1 | - | 1792 | 65 | |||
JV635596.1 | - | 1729 | 65 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
