Assay Details
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047149.1 | NP_001040614.1 | 4 | 664 | 144 | |
XM_015135927.2 | XP_014991413.1 | 1063 | 144 | |||
GenBank mRNA | U19850.1 | - | 4 | 664 | 144 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
