Assay Details
Gene Symbol: CST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV534274.1 | - | 3 | 437 | 98 |
Gene Symbol: RPSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV616428.1 | - | 1126 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
