Assay Details
Gene Symbol: CCL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY288833.1 | - | 1 - 2 | 109 | 95 | |
JV658406.1 | - | 3 - 4 | 354 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
