Assay Details
Gene Symbol: GDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001094714.4 | XP_001094714.2 | 1694 | 86 | ||
XM_015139905.2 | XP_014995391.1 | 818 | 86 | |||
GenBank mRNA | AY288835.1 | - | 1 | 401 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
