Assay Details
Gene Symbol: FASLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032838.1 | NP_001028010.1 | 3 - 4 | 487 | 114 | |
GenBank mRNA | AB035139.1 | - | 3 - 4 | 445 | 114 | |
AF344856.1 | - | 3 - 4 | 487 | 114 | ||
AF530076.1 | - | 3 - 4 | 120 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
