Assay Details
Gene Symbol: CAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033509.2 | NP_001028681.1 | 377 | 59 | ||
GenBank mRNA | AF181954.1 | - | 380 | 59 | ||
JU494369.1 | - | 221 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
