Assay Details
Gene Symbol: ADRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044730.2 | NP_001038195.1 | 1203 | 63 | ||
GenBank mRNA | AF200596.1 | - | 1203 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
