Assay Details
Gene Symbol: CCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032849.1 | NP_001028021.1 | 2 - 3 | 182 | 83 | |
GenBank mRNA | AF449264.1 | - | 2 - 3 | 182 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
