Assay Details
Gene Symbol: CXCL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032934.1 | NP_001028106.1 | 65 | 61 | ||
XM_015146892.2 | XP_015002378.1 | 170 | 61 | |||
XM_015146893.2 | XP_015002379.1 | 168 | 61 | |||
XM_015146894.2 | XP_015002380.1 | 169 | 61 | |||
XM_028853226.1 | XP_028709059.1 | 168 | 61 | |||
GenBank mRNA | AF449283.1 | - | 65 | 61 | ||
JU322651.1 | - | 123 | 61 | |||
JV047889.1 | - | 148 | 61 | |||
JV635595.1 | - | 127 | 61 |

More Information
Set Membership: |
![]() ![]() |
